398209
Lithium tert-butoxide solution
1.0 M in hexanes
Synonym(s):
2-Methyl-2-propanol lithium salt, Lithium 2-methylpropan-2-olate, Lithium tert -butanolate, Lithium tert -butylate, Lithium t -butoxide
About This Item
Recommended Products
form
liquid
reaction suitability
core: lithium
concentration
1.0 M in hexanes
bp
68-70 °C
density
0.69 g/mL at 25 °C
SMILES string
[Li+].CC(C)(C)[O-]
InChI
1S/C4H9O.Li/c1-4(2,3)5;/h1-3H3;/q-1;+1
InChI key
LZWQNOHZMQIFBX-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Aquatic Chronic 2 - Asp. Tox. 1 - Eye Dam. 1 - Flam. Liq. 2 - Repr. 2 - Self-heat. 1 - Skin Corr. 1B - STOT RE 2 Inhalation - STOT SE 3
Target Organs
Central nervous system, Nervous system
Storage Class Code
4.2 - Pyrophoric and self-heating hazardous materials
WGK
WGK 3
Flash Point(F)
-18.4 °F - closed cup
Flash Point(C)
-28 °C - closed cup
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Few Monolayer Atomic Layer Deposition (ALD) on Surfaces and Interfaces for Energy Applications
Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service