394890
N,N-Diethyltrimethylsilylamine
98%
Synonym(s):
(Diethylamino)trimethylsilane, N-(Trimethylsilyl)diethylamine, TMSDEA
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(C2H5)2NSi(CH3)3
CAS Number:
Molecular Weight:
145.32
Beilstein:
635718
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
grade
derivatization grade
vapor density
>1 (vs air)
Assay
98%
refractive index
n20/D 1.411 (lit.)
bp
125-126 °C (lit.)
density
0.767 g/mL at 25 °C (lit.)
SMILES string
CCN(CC)[Si](C)(C)C
InChI
1S/C7H19NSi/c1-6-8(7-2)9(3,4)5/h6-7H2,1-5H3
InChI key
JOOMLFKONHCLCJ-UHFFFAOYSA-N
Looking for similar products? Visit Product Comparison Guide
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Flam. Liq. 2 - Skin Corr. 1B
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
48.2 °F - closed cup
Flash Point(C)
9 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service