Skip to Content
Merck
All Photos(1)

Documents

38215

Sigma-Aldrich

1,4-Diisocyanatobutane

technical, ≥95% (GC)

Synonym(s):

Tetramethylene diisocyanate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
OCN(CH2)4NCO
CAS Number:
Molecular Weight:
140.14
Beilstein:
1756894
MDL number:
UNSPSC Code:
12162002
PubChem Substance ID:

grade

technical

Assay

≥95% (GC)

refractive index

n20/D 1.453

bp

102-104 °C/14 mmHg (lit.)

density

1.105 g/mL at 25 °C (lit.)

storage temp.

2-8°C

SMILES string

O=C=NCCCCN=C=O

InChI

1S/C6H8N2O2/c9-5-7-3-1-2-4-8-6-10/h1-4H2

InChI key

OVBFMUAFNIIQAL-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

replaced by

Product No.
Description
Pricing

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service