Skip to Content
Merck
All Photos(1)

Key Documents

374636

Sigma-Aldrich

Ethyl (S)-2-(trifluoromethylsulfonyloxy)propionate

≥96%

Synonym(s):

Ethyl O-trifluoromethanesulfonyl-L-lactate, Ethyl L-lactate 2-O-triflate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3CH(OSO2CF3)CO2C2H5
CAS Number:
Molecular Weight:
250.19
Beilstein:
3549394
MDL number:
UNSPSC Code:
12352108
PubChem Substance ID:
NACRES:
NA.22

Assay

≥96%

optical activity

[α]20/D −44°, neat

contains

~1% MgO as stabilizer

refractive index

n20/D 1.374 (lit.)

bp

34 °C/0.005 mmHg (lit.)

density

1.342 g/mL at 25 °C (lit.)

functional group

ester
fluoro
triflate

storage temp.

−20°C

SMILES string

CCOC(=O)[C@H](C)OS(=O)(=O)C(F)(F)F

InChI

1S/C6H9F3O5S/c1-3-13-5(10)4(2)14-15(11,12)6(7,8)9/h4H,3H2,1-2H3/t4-/m0/s1

InChI key

RYSBPHXTNVWICH-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Ethyl (S)-2-(trifluoromethylsulfonyloxy)propionate can be used:
  • In the synthesis of tetra-methylated analog of 1,4,7,10-tetraazacyclododecane-1,4,7,10-tetraacetic acid (DOTMA) based chelates for imaging applications.[1]
  • As a substrate in the synthesis of α-silylated carboxyl compounds via Cu-catalyzed stereoinvertive nucleophilic silylation of α-triflyloxy esters.[2]

Pictograms

Corrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Skin Corr. 1B

Storage Class Code

8A - Combustible corrosive hazardous materials

WGK

WGK 3

Flash Point(F)

174.2 °F - closed cup

Flash Point(C)

79 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Properties, solution state behavior, and crystal structures of chelates of DOTMA
Aime S, et al.
Inorganic Chemistry, 50(17), 7955-7965 (2011)
Copper-Catalyzed Substitution of α-Triflyloxy Nitriles and Esters with Silicon Nucleophiles under Inversion of the Configuration
Scharfbier J, et al.
Organic Letters, 19(24), 6562-6565 (2017)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service