373826
Yttrium(III) tris(2,2,6,6-tetramethyl-3,5-heptanedionate)
99.9% trace metals basis
Synonym(s):
Y(TMHD)3
About This Item
Recommended Products
Assay
99.9% trace metals basis
form
solid
reaction suitability
core: yttrium
reagent type: catalyst
mp
173-175 °C
SMILES string
CC(C)(C)C(=O)\C=C(/O[Y](O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)O\C(=C/C(=O)C(C)(C)C)C(C)(C)C)C(C)(C)C
InChI
1S/3C11H20O2.Y/c3*1-10(2,3)8(12)7-9(13)11(4,5)6;/h3*7,12H,1-6H3;/q;;;+3/p-3/b3*8-7-;
InChI key
PPRRRPCEDUWEHL-LWTKGLMZSA-K
Looking for similar products? Visit Product Comparison Guide
Application
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Articles
Nanomaterials are considered a route to the innovations required for large-scale implementation of renewable energy technologies in society to make our life sustainable.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service