Skip to Content
Merck
All Photos(1)

Key Documents

368121

Sigma-Aldrich

1,3-Dibromo-5-fluorobenzene

97%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
Br2C6H3F
CAS Number:
Molecular Weight:
253.89
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

97%

form

liquid

refractive index

n20/D 1.577 (lit.)

bp

204-206 °C/768 mmHg (lit.)

density

2.018 g/mL at 25 °C (lit.)

functional group

bromo
fluoro

SMILES string

Fc1cc(Br)cc(Br)c1

InChI

1S/C6H3Br2F/c7-4-1-5(8)3-6(9)2-4/h1-3H

InChI key

ASWYHZXKFSLNLN-UHFFFAOYSA-N

General description

1,3-Dibromo-5-fluorobenzene is an aryl fluorinated building block.

Application

1,3-Dibromo-5-fluorobenzene may be used in the preparation of ligand 5-fluoro-1,3-di(2-pyridyl)benzene, by a Stille reaction with 2-(tri-nbutylstannyl)pyridine.[1]

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyclometallated platinum (ii) complexes of 1, 3-di (2-pyridyl) benzenes for solution-processable WOLEDs exploiting monomer and excimer phosphorescence.
Mroz W, et al.
Journal of Materials Chemistry, 21(24), 8653-8661 (2011)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service