Skip to Content
Merck
All Photos(1)

Key Documents

365378

Sigma-Aldrich

Cyclopentanone ethylene ketal

97%

Synonym(s):

1,4-Dioxaspiro[4.4]nonane

Sign Into View Organizational & Contract Pricing

Select a Size


Select a Size

Change View

About This Item

Empirical Formula (Hill Notation):
C7H12O2
CAS Number:
Molecular Weight:
128.17
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

refractive index

n20/D 1.448 (lit.)

bp

54-55 °C/35 mmHg (lit.)

density

1.03 g/mL at 25 °C (lit.)

SMILES string

C1CCC2(C1)OCCO2


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service