Skip to Content
Merck
All Photos(1)

Key Documents

345490

Sigma-Aldrich

Zinc p-toluenesulfonate hydrate

Synonym(s):

p-Toluenesulfonic acid zinc salt

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3C6H4SO3)2Zn·xH2O
CAS Number:
Molecular Weight:
407.78 (anhydrous basis)
Beilstein:
4339837
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
powder

Assay

≥98.5% (HPLC)

form

powder

reaction suitability

reagent type: catalyst
core: zinc

concentration

≤15.7% Zn (EDTA titration)

SMILES string

O.[Zn++].Cc1ccc(cc1)S([O-])(=O)=O.Cc2ccc(cc2)S([O-])(=O)=O

InChI

1S/2C7H8O3S.H2O.Zn/c2*1-6-2-4-7(5-3-6)11(8,9)10;;/h2*2-5H,1H3,(H,8,9,10);1H2;/q;;;+2/p-2

InChI key

JCOXFVWERPVZPB-UHFFFAOYSA-L

Looking for similar products? Visit Product Comparison Guide

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Toratane Munegumi et al.
Chirality, 24(2), 188-192 (2011-12-17)
Preferential crystallization of amino acid derivatives by seeding a pure enantiomer into racemic amino acid solutions has been studied for many years. However, few examples of valine derivatives have been reported so far. Although there have been some reports using
Koneni V Sashidhara et al.
Bioorganic & medicinal chemistry letters, 22(12), 3926-3930 (2012-05-23)
First synthesis of novel coumarin-trioxane hybrids is reported. The synthesis was achieved via condensation of β-hydroxyhydroperoxides with coumarinic-aldehydes in presence of p-toluenesulfonic acid in good yields and the novel hybrids were evaluated for their antimalarial activity both in vitro and
G Ozyilmaz et al.
Prikladnaia biokhimiia i mikrobiologiia, 47(2), 217-225 (2011-03-01)
Amperometric glucose biosensors have been developed based on entrapment on platinum (Pt) electrode using cyclic voltammetry technique in glucose oxidase (GOD) and pyrrole containing p-toluenesulfonic acid (pTSA) or sodium p-toluenesulfonate (NapTS) as supporting electrolyte solutions. Both of electrolyte solutions were
Aditya Kulkarni et al.
Organic letters, 13(19), 5124-5127 (2011-09-10)
An environmentally benign procedure for the hydrogenation of unprotected indoles is described. The hydrogenation reaction is catalyzed by Pt/C and activated by p-toluenesulfonic acid in water as a solvent. The efficacy of the method is illustrated by the hydrogenation of
G Shanmugam et al.
Spectrochimica acta. Part A, Molecular and biomolecular spectroscopy, 95, 177-183 (2012-05-29)
Bulk crystals of newly identified organic nonlinear optical material 4-methylanilinium p-toluenesulfonate (PMPT) were grown by slow evaporation solution growth method using ethanol as a solvent. It crystallizes in monoclinic system with a noncentrosymmetric space group P2(1). The formation of the

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service