Skip to Content
Merck
All Photos(1)

Key Documents

316733

Sigma-Aldrich

(1S,2R)-(+)-Ephedrine hemihydrate

98%

Synonym(s):

(1S,2R)-(+)-α-(1-Methylaminoethyl)benzyl alcohol

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
C6H5CH[CH(NHCH3)CH3]OH · 1/2H2O
CAS Number:
Molecular Weight:
174.24
EC Number:
UNSPSC Code:
12352000
PubChem Substance ID:

Assay

98%

optical activity

[α]19/D +40.5°, c = 5 in 1 M HCl

mp

39-43 °C (lit.)

SMILES string

O.CN[C@H](C)[C@@H](O)c1ccccc1.CN[C@H](C)[C@@H](O)c2ccccc2

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Precautionary Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service