All Photos(1)
About This Item
Linear Formula:
BrC6H2F3
CAS Number:
Molecular Weight:
210.98
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
Assay
98%
refractive index
n20/D 1.486 (lit.)
density
1.758 g/mL at 25 °C (lit.)
SMILES string
Fc1cc(F)c(F)c(Br)c1
InChI
1S/C6H2BrF3/c7-4-1-3(8)2-5(9)6(4)10/h1-2H
InChI key
XSMLLZPSNLQCQU-UHFFFAOYSA-N
Signal Word
Warning
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3
Target Organs
Respiratory system
Storage Class Code
3 - Flammable liquids
WGK
WGK 3
Flash Point(F)
89.6 °F - closed cup
Flash Point(C)
32 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Chan Seok Oh et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 25(2), 642-648 (2018-10-20)
Blue thermally activated delayed fluorescent (TADF) devices with short excited-state lifetime, high reverse intersystem crossing rate, and low-efficiency roll-off were developed by managing the molecular structure of donor-acceptor-type blue emitters. Three isomers of blue TADF emitters with a diphenyltriazine acceptor
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service