Skip to Content
Merck
All Photos(1)

Documents

295566

Sigma-Aldrich

Nitric oxide

98.5%

Synonym(s):

Nitrogen monoxide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NO
CAS Number:
Molecular Weight:
30.01
EC Number:
MDL number:
UNSPSC Code:
12142100
PubChem Substance ID:

vapor density

1.05 (vs air)

Quality Level

Assay

98.5%

bp

−151.7 °C (lit.)

mp

−163.6 °C (lit.)

SMILES string

[N]=O

InChI

1S/NO/c1-2

InChI key

MWUXSHHQAYIFBG-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Packaging

Supplied in a carbon steel lecture bottle with a CGA180M/CGA110F needle valve installed.

Compatible with the following:
  • Aldrich® lecture-bottle station systems
  • Aldrich® lecture-bottle gas regulators

Other Notes

Legal Information

Aldrich is a registered trademark of Sigma-Aldrich Co. LLC

hose barb

Product No.
Description
Pricing

purge valve

Product No.
Description
Pricing

Signal Word

Danger

Hazard Statements

Hazard Classifications

Acute Tox. 1 Inhalation - Ox. Gas 1 - Press. Gas Compr. Gas - Skin Corr. 1B

Supplementary Hazards

Storage Class Code

2A - Gases

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Katja Videbaek et al.
Pediatric pulmonology, 54(8), 1182-1189 (2019-04-24)
Primary ciliary dyskinesia (PCD) is a congenital lung disease that leads to recurrent and chronic lung infection. The resulting inflammation causes lung damage and declines in lung function. Mannose-binding lectin (MBL) is a first line host defense protein of importance
Rajeev Malhotra et al.
Arteriosclerosis, thrombosis, and vascular biology, 39(2), 178-187 (2018-12-28)
Objective- Inflammatory stimuli enhance the progression of atherosclerotic disease. Inflammation also increases the expression of hepcidin, a hormonal regulator of iron homeostasis, which decreases intestinal iron absorption, reduces serum iron levels and traps iron within macrophages. The role of macrophage
Kaiyu Zhang et al.
Bioresource technology, 291, 121803-121803 (2019-07-22)
This study investigated the effects of tylosin (0, 10, and 100 mg/kg dry weight) on the denitrification genes and microbial community during the anaerobic digestion of cattle manure. N2 emissions were reduced and N2O emissions were increased by 10 mg/kg tylosin. Adding
C-Y Zhang et al.
European review for medical and pharmacological sciences, 23(14), 6321-6329 (2019-08-01)
To investigate the regulatory effects of simvastatin on the inflammation and oxidative stress in rats with cerebral hemorrhage through the nuclear factor E2-related factor 2-antioxidant response element (Nrf2-ARE) signaling pathway. A total of 120 healthy male rats weighing 280-300 g
Rongjin Wang et al.
Journal of ethnopharmacology, 241, 111969-111969 (2019-05-28)
As a traditional Chinese medicine, Eleutherococcus senticosus (Rupr. & Maxim.) Maxim. leaves (ESL) can treat ischemic, neurasthenia, and hypertension diseases. However, only few studies have been conducted on the mechanism of action of ESL for ischemic disease treatment. This study

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service