Skip to Content
Merck
All Photos(1)

Key Documents

285730

Sigma-Aldrich

Ellipticine

97%

Synonym(s):

5,11-Dimethyl-6H-pyrido[4,3-b]carbazole

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C17H14N2
CAS Number:
Molecular Weight:
246.31
Beilstein:
221300
EC Number:
MDL number:

Assay

97%

SMILES string

Cc1c2ccncc2c(C)c3c4ccccc4[nH]c13

InChI

1S/C17H14N2/c1-10-14-9-18-8-7-12(14)11(2)17-16(10)13-5-3-4-6-15(13)19-17/h3-9,19H,1-2H3

InChI key

CTSPAMFJBXKSOY-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service