255092
mono-Bromoborane methyl sulfide complex
Synonym(s):
Boron monobromide methyl sulfide complex, Bromoborane methyl sulfide complex
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
(CH3)2S·BH2Br
CAS Number:
Molecular Weight:
154.86
MDL number:
UNSPSC Code:
12352112
PubChem Substance ID:
Recommended Products
reaction suitability
reagent type: reductant
density
1.452 g/mL at 25 °C (lit.)
SMILES string
BBr.CSC
InChI
1S/C2H6S.BBrH2/c1-3-2;1-2/h1-2H3;1H2
InChI key
AQRNIOMHNXJPFD-UHFFFAOYSA-N
Application
Reagent for hydroboration and synthesis of bromodialkylboranes.
Signal Word
Danger
Hazard Statements
Precautionary Statements
Hazard Classifications
Eye Dam. 1 - Flam. Liq. 2 - Skin Corr. 1B - Water-react 1
Supplementary Hazards
Storage Class Code
4.3 - Hazardous materials which set free flammable gases upon contact with water
WGK
WGK 3
Flash Point(F)
50.0 °F - closed cup
Flash Point(C)
10 °C - closed cup
Personal Protective Equipment
dust mask type N95 (US), Eyeshields, Gloves
Choose from one of the most recent versions:
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service