Skip to Content
Merck
All Photos(1)

Key Documents

247693

Sigma-Aldrich

2-Chloro-4-fluorotoluene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(Cl)F
CAS Number:
Molecular Weight:
144.57
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
Pricing and availability is not currently available.

Assay

99%

form

liquid

refractive index

n20/D 1.499 (lit.)

bp

154-156 °C (lit.)

density

1.197 g/mL at 25 °C (lit.)

SMILES string

Cc1ccc(F)cc1Cl

InChI

1S/C7H6ClF/c1-5-2-3-6(9)4-7(5)8/h2-4H,1H3

InChI key

CSARJIQZOSVYHA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

2-Chloro-4-fluorotoluene generates vibronically excited, jet-cooled benzyl-type radicals by corona excited supersonic expansion coupled with a pinhole-type glass nozzle technique[1].

Application

2-Chloro-4-fluorotoluene has been used in the preparation of 2-chloro-4-fluoro-benzylbromide[2].

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Flam. Liq. 3 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

3 - Flammable liquids

WGK

WGK 3

Flash Point(F)

122.0 °F - closed cup

Flash Point(C)

50 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chang Soon Huh et al.
The Journal of chemical physics, 136(17), 174306-174306 (2012-05-16)
By means of a technique of corona excited supersonic expansion coupled with a pinhole-type glass nozzle, we generated vibronically excited but jet-cooled benzyl-type radicals from precursor 2-chloro-4-fluorotoluene seeded in a large amount of inert carrier gas He. From an analysis
Synthesis and antitumor activity of platinum (II) complexes containing substituted ethylenediamine ligands.
Brunner H, et al.
European Journal of Medicinal Chemistry, 25(1), 35-44 (1990)

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service