Skip to Content
Merck
All Photos(1)

Key Documents

247057

Sigma-Aldrich

Tetrabutylammonium (meta)periodate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3CH2CH2CH2)4N(IO4)
CAS Number:
Molecular Weight:
433.37
Beilstein:
4621593
EC Number:
MDL number:
UNSPSC Code:
12352302
PubChem Substance ID:
NACRES:
NA.23
form:
solid
Pricing and availability is not currently available.

form

solid

Quality Level

reaction suitability

reagent type: oxidant

concentration

29.3% iodine (titration)

mp

175 °C (dec.) (lit.)

SMILES string

[O-]I(=O)(=O)=O.CCCC[N+](CCCC)(CCCC)CCCC

InChI

1S/C16H36N.HIO4/c1-5-9-13-17(14-10-6-2,15-11-7-3)16-12-8-4;2-1(3,4)5/h5-16H2,1-4H3;(H,2,3,4,5)/q+1;/p-1

InChI key

PYVXLMQALOZKES-UHFFFAOYSA-M

Looking for similar products? Visit Product Comparison Guide

Application

Tetrabutylammonium (meta)periodate (DCM) can be oxidated for the synthesis of aryl nitroso derivatives.[1] It can also be used in the formation of an ionic liquid for the synthesis of aromatic carboxylic acids.[2]

Pictograms

Flame over circleExclamation mark

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Ox. Sol. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Study of Enantioselective Catalysis of Nitroso-Diels-Alder Reaction on Solid Support
Kratka L and Hlavavc J
ChemistrySelect, 2(14), 3987-3992 (2017)
Efficient synthesis of aromatic carboxylic acids from aryl ketones in ionic liquid
Lee JC and Lee J
Synthetic Communications, 36(8), 1071-1074 (2006)
Carmen E Castillo et al.
Chemistry (Weinheim an der Bergstrasse, Germany), 27(15), 4946-4954 (2020-12-23)
Oxoiron(IV) complexes bearing tetradentate ligands have been extensively studied as models for the active oxidants in non-heme iron-dependent enzymes. These species are commonly generated by oxidation of their ferrous precursors. The mechanisms of these reactions have seldom been investigated. In
Michelle L Ingalsbe et al.
Bioorganic & medicinal chemistry letters, 19(17), 4984-4987 (2009-08-08)
Due to the increasing number of strains of drug-resistant bacteria, the development of new antibiotics has become increasingly important. The antibacterial properties of quaternary amines and their derivatives on both Gram-positive and Gram-negative bacteria are well known. However, an encompassing

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service