Skip to Content
Merck
All Photos(3)

Key Documents

229709

Sigma-Aldrich

Iodopentoxide

99.99% trace metals basis

Synonym(s):

Diiodine pentoxide, Iodic anhydride

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
I2O5
CAS Number:
Molecular Weight:
333.81
EC Number:
MDL number:
UNSPSC Code:
12352303
PubChem Substance ID:
To inquire about this product 229709, please contact your local Merck office or dealer. Contact Technical Service

Assay

99.99% trace metals basis

density

5.0 g/mL at 25 °C

SMILES string

O=I(=O)OI(=O)=O

InChI

1S/I2O5/c3-1(4)7-2(5)6

InChI key

BIZCJSDBWZTASZ-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

Flame over circleCorrosion

Signal Word

Danger

Hazard Statements

Hazard Classifications

Eye Dam. 1 - Ox. Sol. 2 - Skin Corr. 1B

Storage Class Code

5.1B - Oxidizing hazardous materials

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service