Skip to Content
Merck
All Photos(1)

Key Documents

197920

Sigma-Aldrich

1-[2-(4-Bromophenoxy)ethyl]pyrrolidine

95%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C12H16BrNO
CAS Number:
Molecular Weight:
270.17
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

95%

form

liquid

refractive index

n20/D 1.557 (lit.)

bp

138-140 °C/1.2 mmHg (lit.)

density

1.304 g/mL at 25 °C (lit.)

functional group

bromo

SMILES string

Brc1ccc(OCCN2CCCC2)cc1

InChI

1S/C12H16BrNO/c13-11-3-5-12(6-4-11)15-10-9-14-7-1-2-8-14/h3-6H,1-2,7-10H2

InChI key

YDWKSSWZGXRQET-UHFFFAOYSA-N

Application

1-[2-(4-Bromophenoxy)ethyl]pyrrolidine was used in the synthesis of 4-OCH2CH2 -pyrrolidine[1].

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Chuansheng Niu et al.
Bioorganic & medicinal chemistry letters, 19(20), 5829-5832 (2009-09-19)
A series of 5-vinyl-3-pyridinecarbonitriles were synthesized and evaluated as PKCtheta inhibitors. The systematic optimization of 4-[(4-methyl-1H-indol-5-yl)amino]-5-[(E)-2-phenylvinyl]-3-pyridinecarbonitrile 3 resulted in the identification of compound 23e as a potent PKCtheta inhibitor with good selectivity over PKCdelta.

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service