Skip to Content
Merck
All Photos(1)

Key Documents

191639

Sigma-Aldrich

2,4,6-Trimethylbenzyl alcohol

99%

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
(CH3)3C6H2CH2OH
CAS Number:
Molecular Weight:
150.22
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

Assay

99%

form

solid

bp

140 °C/15 mmHg (lit.)

mp

87-89 °C (lit.)

functional group

hydroxyl

SMILES string

Cc1cc(C)c(CO)c(C)c1

InChI

1S/C10H14O/c1-7-4-8(2)10(6-11)9(3)5-7/h4-5,11H,6H2,1-3H3

InChI key

LODDFDHPSIYCTK-UHFFFAOYSA-N

General description

2,4,6-Trimethylbenzyl alcohol undergoes oxidation in the presence of molecular iodine and potassium carbonate in 2,2,2-trifluoroethanol.

Application

2,4,6-Trimethylbenzyl alcohol was used in the synthesis of 3-(2,4,6-trimethylbenzyl)indole.

Storage Class Code

11 - Combustible Solids

WGK

WGK 2

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

H G Shertzer et al.
Archives of toxicology, 70(12), 830-834 (1996-01-01)
Many dietary constituents, such as indole-3-carbinol, are chemoprotective in toxicity and carcinogenicity bioassays. Indole-3-carbinol and related congeners appear to protect partly via radical and electrophile scavenging. To develop better chemoprotective indoles with lower intrinsic toxicity, we performed molecular graphic and
Facile oxidative conversion of alcohols to esters using molecular iodine.
Mori N and Togo H.
Tetrahedron, 61(24), 5915-5925 (2005)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service