Skip to Content
Merck
All Photos(1)

Documents

183911

Sigma-Aldrich

Methyl 2-chloroacetoacetate

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3COCH(Cl)CO2CH3
CAS Number:
Molecular Weight:
150.56
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

form

liquid

refractive index

n20/D 1.446 (lit.)

bp

137 °C (lit.)

mp

−33-−32.7 °C (lit.)

density

1.236 g/mL at 25 °C (lit.)

SMILES string

COC(=O)C(Cl)C(C)=O

InChI

1S/C5H7ClO3/c1-3(7)4(6)5(8)9-2/h4H,1-2H3

InChI key

GYQRIAVRKLRQKP-UHFFFAOYSA-N

Application

Methyl 2-chloroacetoacetate was used in one-pot synthesis of 2-arylimidazole-4-carboxylic acids. It was also used in synthesis of 3,5-disubstituted 1,2,4-triazoles.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

161.6 °F - closed cup

Flash Point(C)

72 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Joshua C Yoburn et al.
Organic letters, 7(17), 3801-3803 (2005-08-12)
A versatile, one-pot synthesis of 2-arylimidazole-4-carboxylic acids from arylamidines and methyl-2-chloroacetoacetate is described. The transformation is chemoselective, and reaction conditions are mild. Moreover, the flexibility of the strategy offers rapid access to two important classes of biaryl compounds, both 2-arylimidazoles
'One-flask'synthesis to 3, 5-disubstituted 1, 2, 4-triazoles from aldehydes with hydrazonoyl hydrochlorides via 1, 3-dipolar cycloaddition.
Tseng W-C, et al.
Tetrahedron, 67(29), 5339-5345 (2011)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service