Skip to Content
Merck
All Photos(1)

Documents

180734

Sigma-Aldrich

3,4-Dihydro-2-methoxy-2H-pyran

99%

Synonym(s):

2-Methoxy-3,4-dihydro-2H-pyran

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H10O2
CAS Number:
Molecular Weight:
114.14
Beilstein:
107274
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

99%

refractive index

n20/D 1.442 (lit.)

density

1.006 g/mL at 25 °C (lit.)

SMILES string

COC1CCC=CO1

InChI

1S/C6H10O2/c1-7-6-4-2-3-5-8-6/h3,5-6H,2,4H2,1H3

InChI key

XCYWUZHUTJDTGS-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Pictograms

FlameExclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Eye Irrit. 2 - Flam. Liq. 3

Storage Class Code

3 - Flammable liquids

WGK

WGK 1

Flash Point(F)

78.8 °F - closed cup

Flash Point(C)

26 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

B Ballantyne et al.
Human toxicology, 8(3), 229-235 (1989-05-01)
1. The LC50 values for acrolein (AC) vapour to Sprague-Dawley rats (combined sexes) were determined to be 26 ppm (1 h) and 8.3 ppm (4 h). Signs of severe irritancy were present, and death was due to lung injury. 2.
The acute toxicity of methoxydihydropyran.
D E Dodd et al.
Veterinary and human toxicology, 30(6), 545-550 (1988-12-01)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service