Skip to Content
Merck
All Photos(1)

Key Documents

17520

Sigma-Aldrich

4-Bromo-DL-mandelic acid

technical, ≥90% (HPLC)

Synonym(s):

4-Bromo-α-hydroxyphenylacetic acid

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH(OH)CO2H
CAS Number:
Molecular Weight:
231.04
Beilstein:
1567909
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22

grade

technical

Assay

≥90% (HPLC)

mp

116-118 °C (lit.)

functional group

bromo
carboxylic acid
hydroxyl

SMILES string

OC(C(O)=O)c1ccc(Br)cc1

InChI

1S/C8H7BrO3/c9-6-3-1-5(2-4-6)7(10)8(11)12/h1-4,7,10H,(H,11,12)

InChI key

BHZBRPQOYFDTAB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application

4-Bromo-DL-mandelic acid (4-Bromo-α-hydroxyphenylacetic acid) may be used in the synthesis of (.+-.)-methyl 4-bromo-α-hydroxyphenylacetate.

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service