Skip to Content
Merck
All Photos(1)

Key Documents

159581

Sigma-Aldrich

3-Cyanopropionaldehyde dimethyl acetal

98%

Synonym(s):

4,4-Dimethoxybutyronitrile

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NCCH2CH2CH(OCH3)2
CAS Number:
Molecular Weight:
129.16
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Quality Level

Assay

98%

form

liquid

refractive index

n20/D 1.419 (lit.)

bp

90-91 °C/14 mmHg (lit.)

density

0.992 g/mL at 25 °C (lit.)

SMILES string

COC(CCC#N)OC

InChI

1S/C6H11NO2/c1-8-6(9-2)4-3-5-7/h6H,3-4H2,1-2H3

InChI key

XJHNCGVHPLDMRK-UHFFFAOYSA-N

Application

3-Cyanopropionaldehyde dimethyl acetal was used in the synthesis of 7-methyl-2-naphthalenecarbonitrile, precursor of the anticoagulant agents YM-60828 or YM-96765.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

192.2 °F - closed cup

Flash Point(C)

89 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Customers Also Viewed

Hiroyuki Koshio et al.
Chemical & pharmaceutical bulletin, 53(4), 448-450 (2005-04-02)
A practical and cost-effective procedure has been developed for the synthesis of 7-methyl-2-naphthalenecarbonitrile, the precursor of the anticoagulant agents YM-60828 or YM-96765. This new route generates the key intermediate in only two steps from readily available 3-cyanopropionaldehyde dimethyl acetal and

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service