Skip to Content
Merck
All Photos(1)

Key Documents

138347

Sigma-Aldrich

1-Bromo-3-nitrobenzene

99%

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C6H4BrNO2
CAS Number:
Molecular Weight:
202.01
Beilstein:
2043212
EC Number:
MDL number:
NACRES:
NA.24

Assay

99%

SMILES string

[O-][N+](=O)c1cccc(Br)c1

InChI

1S/C6H4BrNO2/c7-5-2-1-3-6(4-5)8(9)10/h1-4H

InChI key

FWIROFMBWVMWLB-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

230.0 °F - closed cup

Flash Point(C)

110 °C - closed cup


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service