Skip to Content
Merck
All Photos(1)

Key Documents

131547

Sigma-Aldrich

L-Proline

≥99%

Synonym(s):

(S)-Pyrrolidine-2-carboxylic acid

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C5H9NO2
CAS Number:
Molecular Weight:
115.13
Beilstein:
80810
EC Number:
MDL number:
UNSPSC Code:
12352103

Assay

≥99%

optical activity

[α]20/D −84°, c = 4 in H2O

optical purity

ee: 98% (GLC)

mp

228 °C (dec.) (lit.)

SMILES string

OC(=O)[C@@H]1CCCN1

InChI

1S/C5H9NO2/c7-5(8)4-2-1-3-6-4/h4,6H,1-3H2,(H,7,8)/t4-/m0/s1

InChI key

ONIBWKKTOPOVIA-BYPYZUCNSA-N

Looking for similar products? Visit Product Comparison Guide

Application

Catalyst for enantioselective reduction of ketones. Ligand used in TLC resolution of amino acids. Synthon for dolaproine and related diastereomers.

replaced by

Product No.
Description
Pricing

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Journal of Planar Chromatography--Modern TLC, 7, 126-126 (1994)
Tetrahedron Asymmetry, 4, 2255-2255 (1993)
The Journal of Organic Chemistry, 59, 6287-6287 (1994)

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service