Skip to Content
Merck
All Photos(1)

Key Documents

125148

Sigma-Aldrich

2-Chloro-4-nitrotoluene

97%

Synonym(s):

2-Chloro-1-methyl-4-nitrobenzene

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
CH3C6H3(NO2)Cl
CAS Number:
Molecular Weight:
171.58
Beilstein:
1817924
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:

Assay

97%

mp

60-64 °C (lit.)

solubility

acetonitrile: soluble

SMILES string

Cc1ccc(cc1Cl)[N+]([O-])=O

InChI

1S/C7H6ClNO2/c1-5-2-3-6(9(10)11)4-7(5)8/h2-4H,1H3

InChI key

LLYXJBROWQDVMI-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

FT-IR and FT-Raman spectra of 2-chloro-4-nitrotoluene have been studied in the regions 4000-400 and 4000-100 cm-1.

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

6.1C - Combustible acute toxic Cat.3 / toxic compounds or compounds which causing chronic effects

WGK

WGK 2

Flash Point(F)

212.0 °F - closed cup

Flash Point(C)

100 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Combined spectroscopic and DFT studies on 2-chloro-4-nitrotoluene and 4-chloro-2-nitrotoluene.
Arjunan V, et al.
Journal of Molecular Structure, 10(16), 82-96 (2012)
Gundula Wolf et al.
Journal of biotechnology, 128(4), 801-812 (2007-02-13)
An improved method for deconvoluting complex spectral maps from bidimensional fluorescence monitoring is presented, relying on a combination of principal component analysis (PCA) and feedforward artificial neural networks (ANN). With the aim of reducing ANN complexity, spectral maps are first
[Method of determining p-nitrotoluene, 2-chloro-4-nitrotoluene and 3-chloro-4-methylaniline in the air].
I I Pilenkova et al.
Gigiena truda i professional'nye zabolevaniia, (1)(1), 43-44 (1980-01-01)
G Wolf et al.
Biotechnology and bioengineering, 72(3), 297-306 (2001-01-03)
The use of two-dimensional scanning fluorometry as an on-line, noninvasive, in situ bioreactor monitoring technique is extended to complex bioprocesses using mixed cultures, with particular attention to biofilm systems. Using the example of spectra subtraction, it is demonstrated that established

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service