Skip to Content
Merck
All Photos(1)

Key Documents

124664

Sigma-Aldrich

4-Bromophenylacetonitrile

98%

Synonym(s):

4-Bromobenzyl cyanide

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
BrC6H4CH2CN
CAS Number:
Molecular Weight:
196.04
EC Number:
MDL number:
UNSPSC Code:
12352100
PubChem Substance ID:
NACRES:
NA.22
form:
solid
Assay:
98%
To inquire about this product 124664, please contact your local Merck office or dealer. Contact Technical Service

Assay

98%

form

solid

mp

47-49 °C (lit.)

functional group

bromo
nitrile

SMILES string

Brc1ccc(CC#N)cc1

InChI

1S/C8H6BrN/c9-8-3-1-7(2-4-8)5-6-10/h1-4H,5H2

InChI key

MFHFWRBXPQDZSA-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

General description

4-Bromophenylacetonitrile undergoes reduction using 1,1,3,3-tetramethyldisiloxane activated by a non-toxic titanium(IV) isopropoxide to yield 4-bromophenylethylamine[1].

Application

4-Bromophenylacetonitrile has been used in the synthesis of :
  • new fluorene-based alternating polymers[2]
  • light-emitting copolymer, poly(N 4,N 4-bis-(4-phenyl)-N 4,N 4′-bis-[4-(2-ethylhexyloxy)phenyl]-biphenyl-4,4′-diamine-co-1,2-bis(4′-phenyl)-1-(9′′,9′′-dihexyl-3-fluorenyl)ethane) using a Yamamoto coupling reaction[3]
  • N-methyl-3,4-bis(4-(N-(1-naphthyl)phenylamino)phenyl)maleimide, a novel non-doped red organic light-emitting diode (OLED) material[4]

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Classifications

Acute Tox. 4 Dermal - Acute Tox. 4 Inhalation - Acute Tox. 4 Oral - Aquatic Chronic 3 - Eye Irrit. 2 - Skin Irrit. 2 - STOT SE 3

Target Organs

Respiratory system

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

235.4 °F - closed cup

Flash Point(C)

113 °C - closed cup

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Hsiu-Chih Yeh et al.
Chemical communications (Cambridge, England), (20)(20), 2632-2633 (2003-11-05)
Bright (maximum 10034 cd m(-2), 455 cd m(-2) at 20 mA cm(-2)) and efficient (maximum 2.4% at 4 mA cm(-2)) red (lambda(max)el 634-636 nm) organic light-emitting diodes employ arylamino-substituted fumaronitrile as the novel host emitter, which is readily prepared and
Synthesis and device performance of new efficient blue copolymers containing biphenylenevinylene and triphenyldiamine.
Oh DH, et al.
Macromolecular Research, 19(6), 629-634 (2011)
Polymeric light emitting properties and structural relationships of fluorene-based conjugated copolymers containing various hole transporting derivatives.
Park JH, et al.
Organic Electronics, 8(2), 272-285 (2007)
A mild and efficient method for the reduction of nitriles.
Laval S, et al.
Tetrahedron Letters, 50(50), 7005-7007 (2009)
Bocheng Wu et al.
Bioorganic & medicinal chemistry, 28(6), 115345-115345 (2020-02-18)
Signal transducer and activator of transcription 3 (STAT3) is an oncogenic transcription factor which has been recognized as a promising cancer therapeutic target. Small molecule pyrimethamine (PYM) is a known direct inhibitor of activated STAT3 and it is currently under

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service