Merck
Peptide gel in a scaffold as a composite matrix for endothelial cells.
Journal of biomedical materials research. Part A (2015-03-27)
Cristina Martínez-Ramos, María Arnal-Pastor, Ana Vallés-Lluch, Manuel Monleón Pradas
ABSTRACT
The performance of a composite environment with human umbilical vein endothelial cells (HUVECs) has been studied to provide an in vitro proof of concept of their potential of being easily vascularized. These cells were seeded in 1 mm thick scaffolds whose pores had been filled with a self-assembling peptide gel, seeking to improve cell adhesion, and viability of these very sensitive cells. The combination of the synthetic elastomer poly(ethyl acrylate), PEA, scaffold and the RAD16-I peptide gel provides cells with a friendly ECM-like environment inside a mechanically resistant structure. Immunocytochemistry, flow cytometry and scanning electron microscopy were used to evaluate the cell cultures. The presence of the self-assembling peptide filling the pores of the scaffolds resulted in a truly 3D nanoscale context mimicking the extracellular matrix environment, and led to increased cells survival, proliferation as well as developed cell-cell contacts. The combined system consisting of PEA scaffolds and RAD16-I, is a very interesting approach as seems to enhance endothelization, which is the first milestone to achieve vascularized constructs.
Sigma-Aldrich
Benzoin, purified by sublimation, ≥99.5%
Sigma-Aldrich
Hydrocortisone, meets USP testing specifications
Sigma-Aldrich
Hydrocortisone, γ-irradiated, powder, BioXtra, suitable for cell culture
Sigma-Aldrich
Hydrocortisone, ≥98% (HPLC)
Sigma-Aldrich
Hydrocortisone, BioReagent, suitable for cell culture
Sigma-Aldrich
2-Hydroxyethyl acrylate, 96%, contains 200-650 ppm monomethyl ether hydroquinone as inhibitor
Sigma-Aldrich
Ethyl acrylate, contains 10-20 ppm MEHQ as inhibitor, 99%
Sigma-Aldrich
Benzoin, 98%
Sigma-Aldrich
Ethyl acrylate, ≥99.5%, stabilized
Sigma-Aldrich
Phenol Red, powder, BioReagent, suitable for cell culture
Sigma-Aldrich
DAPI, for nucleic acid staining
Sigma-Aldrich
Phenol Red, ACS reagent