Merck
  • The leukocyte common antigen (CD45) on human pre-B leukemia cells: variant glycoprotein form expression during the cell exposure to phorbol ester is blocked by a nonselective protein kinase inhibitor H7.
The leukocyte common antigen (CD45) on human pre-B leukemia cells: variant glycoprotein form expression during the cell exposure to phorbol ester is blocked by a nonselective protein kinase inhibitor H7.
Neoplasma (1997-01-01)
J Duraj, J Sedlák, P Rauko, B Chorváth
ABSTRACT
The human pre-B acute lymphoblastic leukemia cell line REH6 was utilized for characterization of CD45 glycoprotein by monoclonal antibodies (mAb) recognizing four distinct CD45 antigen specificities, i.e. nonrestricted CD45, restricted CD45RA, CD45RB and CD45R0. Immunoprecipitation revealed two antigen specificities on REH6 cells of m.w. 220 kDa and 190 kDa, both presenting wide range of isoelectric point pI approximately 6.0-7.5. Nonrestricted CD45 epitopes were not affected by the sialyl acid cleavage with sodium metaperiodate or neuraminidase, but were sensitive to both, tunicamycin, the N-glycosylation inhibitor and monensin, an inhibitor of protein transport through the Golgi compartment. O-sialoglycoprotein endopeptidase from Pasteurella haemolytica A1 partially cleaved CD45RA and CD45RB epitopes, while nonrestricted CD45 determinants were not affected by this enzyme. Limited proteolysis of this antigen resulted in the appearance of 160-180 kDa peptide domains which retained CD45 epitopes. Further, the treatment of cells with phorbol myristate acetate (PMA) induced marked down-regulation of 220 and 190 kDa isoforms and the appearance of new 210, 180 and 170 kDa variant glycoprotein forms which were not found on parental cells. This PMA effect was not accompanied by the programmed cell death and was markedly blocked by a nonselective protein kinase (PK) inhibitor isoquinoline sulfonamide H7. Modulation of CD45 by phorbol esters might serve as an in vitro model for an additional insight into the function of CD45 in hematopoietic cells.
Sigma-Aldrich
1-(5-Isoquinolinesulfonyl)-3-methylpiperazine, ≥98.0% (HPLC)