Merck
  • Occupational allergic contact dermatitis caused by epoxy diacrylate in ultraviolet-light-cured paint, and bisphenol A in dental composite resin.
Occupational allergic contact dermatitis caused by epoxy diacrylate in ultraviolet-light-cured paint, and bisphenol A in dental composite resin.
Contact dermatitis (1995-08-01)
R Jolanki, L Kanerva, T Estlander
ABSTRACT
Allergic contact dermatitis (ACD) caused by epoxy di(meth)acrylates or bisphenol A is rare. Here 2 such cases are reported. A dental assistant had allergic contact dermatitis (ACD) caused by bisphenol A contained in dental composite resin (DCR) products based on epoxy dimethacrylate. The contact allergy was verified by allergic patch test reactions to bisphenol A and 2 DCRs. The DCRs giving allergic reactions were analyzed, and 0.014-0.015% of bisphenol A was detected. Occupational ACD caused by bisphenol A in dental composite resins has not been described before. The other patient was a male process worker in a paint factory. He was sensitized by an epoxy diacrylate, 2,2-bis[4-(2-hydroxy-3-acryloxypropoxy)phenyl]-propane (BIS-GA), and other acrylate compounds contained in raw materials of ultraviolet-light-curable paint. The epoxy diacrylate gave an allergic patch test reaction down to 0.016% in pet. He also had an allergic patch test reaction to several other acrylate compounds, 2-hydroxyethyl acrylate, 2-hydroxypropyl acrylate, 1,4-butanediol diacrylate, 1,6-hexanediol diacrylate, diethyleneglycol diacrylate, triethylene glycol diacrylate, and tripropylene glycol diacrylate, indicating cross and/or concomitant sensitization.
Sigma-Aldrich
Bisphenol A glycerolate (1 glycerol/phenol) diacrylate, contains MEHQ as inhibitor