Merck
  • Transparent high-performance thin film transistors from solution-processed SnO2 /ZrO2 gel-like precursors.
Transparent high-performance thin film transistors from solution-processed SnO2 /ZrO2 gel-like precursors.
Advanced materials (Deerfield Beach, Fla.) (2012-11-20)
Jaewon Jang, Rungrot Kitsomboonloha, Sarah L Swisher, Eung Seok Park, Hongki Kang, Vivek Subramanian
ABSTRACT
This work employs novel SnO(2) gel-like precursors in conjunction with sol-gel deposited ZrO(2) gate dielectrics to realize high-performance transparent transistors. Representative devices show excellent performance and transparency, and deliver mobility of 103 cm(2) V(-1) s(-1) in saturation at operation voltages as low as 2 V, a sub-threshold swing of only 0.3 V/decade, and /(on) //(off) of 10(4) ~10(5) .
Sigma-Aldrich
Tin(IV) oxide, ≥99.99% trace metals basis
Sigma-Aldrich
Tin(IV) oxide, nanopowder, ≤100 nm avg. part. size
Sigma-Aldrich
Tin(IV) oxide, −325 mesh, 99.9% trace metals basis