MilliporeSigma
  • Synthetic access to poly-substituted 6-alkoxyindoles from 1,3-cyclohexanediones and nitroolefins through facile aromatization reaction.
Synthetic access to poly-substituted 6-alkoxyindoles from 1,3-cyclohexanediones and nitroolefins through facile aromatization reaction.
The Journal of organic chemistry (2009-11-10)
Li-Jian Ma, Xiao-Xia Li, Tomoyo Kusuyama, Ibrahim El-Tantawy El-Sayed, Tsutomu Inokuchi
ABSTRACT
6-Alkoxy-3-arylindoles were efficiently prepared from 1,3-cyclohexanedione enol ethers and beta-nitrostyrenes. Michael addition using the kinetically generated enolate, followed by Zn reduction of a nitro group of the resulting adducts produced the nitrones which were then treated with acetic anhydride to induce aromatization by dehydration and N-acetylation, which furnished the desired indoles by the DDQ oxidation. This methodology provides an easy entry toward various poly-substituted 6-alkoxyindoles.
Sigma-Aldrich
1,3-Cyclohexanedione, 97%