Merck
New Pd-catalyzed selective reduction of carboxylic acids to aldehydes.
Chemical communications (Cambridge, England) (2002-07-19)
Lukas J Goossen, Keya Ghosh
ABSTRACT
A catalyst generated in situ from palladium acetate and tricyclohexylphosphine efficiently catalyzes the reduction of carboxylic acids with sodium hypophosphite in the presence of pivalic anhydride to give aldehydes with high selectivity. The low cost and convenient handling of the reagents makes this process a valuable alternative to hydrogenations and metal hydride reductions.
Sigma-Aldrich
Sodium hypophosphite monohydrate, ≥99%
Sigma-Aldrich
Sodium hypophosphite, puriss., meets analytical specification of BPC63, anhydrous, 98-101%