MilliporeSigma
  • Novel amyloid-beta specific scFv and VH antibody fragments from human and mouse phage display antibody libraries.
Novel amyloid-beta specific scFv and VH antibody fragments from human and mouse phage display antibody libraries.
Journal of neuroimmunology (2010-05-11)
M Medecigo, K Manoutcharian, V Vasilevko, T Govezensky, M E Munguia, B Becerril, A Luz-Madrigal, L Vaca, D H Cribbs, G Gevorkian
ABSTRACT
Anti-amyloid immunotherapy has been proposed as an appropriate therapeutic approach for Alzheimer's disease (AD). Significant efforts have been made towards the generation and assessment of antibody-based reagents capable of preventing and clearing amyloid aggregates as well as preventing their synaptotoxic effects. In this study, we selected a novel set of human anti-amyloid-beta peptide 1-42 (Abeta1-42) recombinant monoclonal antibodies in a single chain fragment variable (scFv) and a single-domain (VH) format. We demonstrated that these antibody fragments recognize in a specific manner amyloid-beta deposits in APP/Tg mouse brains, inhibit toxicity of oligomeric Abeta1-42 in neuroblastoma cell cultures in a concentration-dependent manner and reduced amyloid deposits in APP/Tg2576 mice after intracranial administration. These antibody fragments recognize epitopes in the middle/C-terminus region of Abeta, which makes them strong therapeutic candidates due to the fact that most of the Abeta species found in the brains of AD patients display extensive N-terminus truncations/modifications.
Sigma-Aldrich
Anti-β-Amyloid (13-28) antibody, Mouse monoclonal, clone BAM90.1, purified from hybridoma cell culture