Merck
  • Synthesis of fatty acyl CoA and other thiol esters using N-hydroxysuccinimide esters of fatty acids.
Synthesis of fatty acyl CoA and other thiol esters using N-hydroxysuccinimide esters of fatty acids.
Journal of lipid research (1969-05-01)
A Al-Arif, M Blecher
ABSTRACT
N-Hydroxysuccinimide esters of long-chain fatty acids have been used to synthesize the CoA and thioglycolic acid thiol esters of palmitic and 3-ketopalmitic acids in high yield and with a minimum of untoward side reactions.
Sigma-Aldrich
Palmitic acid N-hydroxysuccinimide ester, ≥98% (TLC)
Sign Into View Organizational & Contract Pricing
SKUPack SizeAvailabilityPriceQuantity
1 g
Estimated to ship on April 19, 2025
$136.00
5 g
Estimated to ship on April 19, 2025
$497.00