Skip to Content
Merck
All Photos(2)

Key Documents

V900222

Sigma-Aldrich

Ammonium chloride

Vetec, reagent grade, 99%

Synonym(s):

Salmiac

Sign Into View Organizational & Contract Pricing


About This Item

Linear Formula:
NH4Cl
CAS Number:
Molecular Weight:
53.49
Beilstein:
4371014
EC Number:
MDL number:
UNSPSC Code:
12352300
PubChem Substance ID:
grade:
reagent grade
form:
powder

grade

reagent grade

vapor density

1.9 (vs air)

vapor pressure

1 mmHg ( 160.4 °C)

product line

Vetec

Assay

99%

form

powder

pH

4.5-5.5 (20 °C, 50 g/L)

mp

340 °C (subl.) (lit.)

SMILES string

N.Cl

InChI

1S/ClH.H3N/h1H;1H3

InChI key

NLXLAEXVIDQMFP-UHFFFAOYSA-N

Looking for similar products? Visit Product Comparison Guide

Application



  • Ammonium chloride catalyzed Knoevenagel condensation in PEG-400 as ecofriendly solvent.: The research demonstrates the use of ammonium chloride as a catalyst for the Knoevenagel condensation reaction in an ecofriendly solvent (PEG-400). This highlights the potential for sustainable chemical processes using ammonium chloride (Indian Journal of Chemistry -Section B, 2021).

  • Recovery of Vanadium by ammonium chloride precipitation method using response surface methodology.: This study investigates the recovery of vanadium using ammonium chloride precipitation. The results show that a high recovery rate of over 90% can be achieved, demonstrating the efficacy of ammonium chloride in metal recovery processes (Gharagozlou et al., 2021).


  • Ammonium Escorted Chloride Chemistry in Stabilizing Aqueous Chloride Ion Battery.: This paper discusses the stabilization of aqueous chloride ion batteries using ammonium chloride, which enhances battery performance and longevity. The research provides insights into the development of advanced energy storage systems (Sun et al., 2022).



Legal Information

Vetec is a trademark of Merck KGaA, Darmstadt, Germany

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral - Eye Irrit. 2

Storage Class Code

13 - Non Combustible Solids

WGK

WGK 1

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Cyrill A Rentsch et al.
European urology, 66(4), 677-688 (2014-03-29)
Whether the commonly used bacillus Calmette-Guérin (BCG) strains Connaught and Tice confer different treatment responses in non-muscle-invasive bladder cancer (NMIBC) is unknown. To compare clinical efficacy, immunogenicity, and genetics of BCG Connaught and Tice. A prospective randomized single-institution trial with
Chloé Lescale et al.
FASEB journal : official publication of the Federation of American Societies for Experimental Biology, 29(2), 455-463 (2014-11-08)
Within the bone marrow, the endosteal niche plays a crucial role in B-cell differentiation. Because spaceflight is associated with osteoporosis, we investigated whether changes in bone microstructure induced by a ground-based model of spaceflight, hind limb unloading (HU), could affect
Allison L Boyd et al.
The Journal of experimental medicine, 211(10), 1925-1935 (2014-09-03)
Allogeneic hematopoietic stem cell (HSC) transplantation (HSCT) is currently the leading strategy to manage acute myeloid leukemia (AML). However, treatment-related morbidity limits the patient generalizability of HSCT use, and the survival of leukemic stem cells (LSCs) within protective areas of
Christine Burkard et al.
PloS one, 9(7), e101762-e101762 (2014-07-16)
Studies of viral entry into host cells often rely on the detection of post-entry parameters, such as viral replication or the expression of a reporter gene, rather than on measuring entry per se. The lack of assays to easily detect
Sewa Rijal et al.
Blood, 125(18), 2815-2824 (2015-03-05)
Phosphoinositide signaling regulates diverse cellular functions. Phosphoinositide-3 kinase (PI3K) generates PtdIns(3,4,5)P3 and PtdIns(3,4)P2, leading to the activation of proliferative and anti-apoptotic signaling pathways. Termination of phosphoinositide signaling requires hydrolysis of inositol ring phosphate groups through the actions of PtdIns(3,4,5)P3 3-phosphatase

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service