SAAR5822320
Sodium chloride
univAR™
Synonym(s):
Halite, NaCl
Sign Into View Organizational & Contract Pricing
All Photos(1)
About This Item
Linear Formula:
NaCl
CAS Number:
Molecular Weight:
58.44
Beilstein:
3534976
MDL number:
UNSPSC Code:
12352302
grade:
reagent grade
Recommended Products
grade
reagent grade
product line
univAR™
packaging
bucket of 5 kg (SAAR5822320FL)
poly bottle of 1 × 500 g (SAAR5822320EM)
mp
801 °C (lit.)
storage temp.
2-30°C
SMILES string
[Na+].[Cl-]
InChI
1S/ClH.Na/h1H;/q;+1/p-1
InChI key
FAPWRFPIFSIZLT-UHFFFAOYSA-M
Looking for similar products? Visit Product Comparison Guide
Application
Analysed reagent
For laboratory use only
Legal Information
univAR is a trademark of Merck KGaA, Darmstadt, Germany
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Lot/Batch Number
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service