Skip to Content
Merck
All Photos(1)

Documents

PHL89280

Silybin (A + B mixture)

phyproof® Reference Substance

Synonym(s):

Silybin, (2R,3R)-3,5,7-Trihydroxy-2-[3-(4-hydroxy-3-methoxyphenyl)-2-hydroxymethyl-2,3-dihydrobenzo[1,4]dioxin-6-yl]chroman-4-one, Silibinin

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H22O10
CAS Number:
Molecular Weight:
482.44
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥95.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages

format

neat

storage temp.

2-8°C

SMILES string

COc1cc(ccc1O)C2Oc3cc(ccc3OC2CO)[C@H]4Oc5cc(O)cc(O)c5C(=O)[C@@H]4O

InChI

1S/C25H22O10/c1-32-17-6-11(2-4-14(17)28)24-20(10-26)33-16-5-3-12(7-18(16)34-24)25-23(31)22(30)21-15(29)8-13(27)9-19(21)35-25/h2-9,20,23-29,31H,10H2,1H3/t20?,23-,24?,25+/m0/s1

InChI key

SEBFKMXJBCUCAI-DBMPWETRSA-N

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application


  • Opioid receptor binding studies: An analysis of irreversible binding of cis-(+)-3-methylfentanyl isothiocyanate to the delta opioid receptor demonstrates its unique interaction and potential for understanding opioid receptor pharmacology (Zhu et al., 1996).

  • Forensic detection of synthetic opioids: A forensic study details the screening and quantification of opioid drugs, including cis-3-Methylfentanyl, in post-mortem samples using advanced mass spectrometry techniques, crucial for toxicological analysis and forensic applications (Gergov et al., 2009).

  • Molecular docking studies on fentanyl: A study featuring molecular docking to reveal a novel binding site model for fentanyl on the mu-opioid receptor contributes valuable insights that are applicable to cis-3-Methylfentanyl′s interaction at opioid receptors, useful for developing targeted analgesics (Subramanian et al., 2000).



Other Notes

This compound is commonly found in plants of the genus: silybum

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Pictograms

Exclamation mark

Signal Word

Warning

Hazard Statements

Hazard Classifications

Acute Tox. 4 Oral

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service