Skip to Content
Merck
All Photos(1)

Documents

PHL86755

Colupulone (stable DCHA salt)

phyproof® Reference Substance

Synonym(s):

Colupulone (dicyclohexylammonium) salt

Sign Into View Organizational & Contract Pricing


About This Item

Empirical Formula (Hill Notation):
C25H36O4·C12H23N
CAS Number:
Molecular Weight:
581.87
UNSPSC Code:
41116107
NACRES:
NA.24

grade

primary reference standard

product line

phyproof® Reference Substance

Assay

≥90.0% (HPLC)

manufacturer/tradename

PhytoLab

application(s)

food and beverages
pharmaceutical

storage temp.

−20°C

General description

This substance is a primary reference substance with assigned absolute purity (considering chromatographic purity, water, residual solvents, inorganic impurities). The exact value can be found on the certificate. Produced by PhytoLab GmbH & Co. KG

Application

The counterion is determined quantitatively and treated as an impurity in the calculation of the absolute purity of this reference substance. The assigned absolute purity on the certificate of analysis of this reference standard, therefore, refers to the pure colupulone. Please note that the given CAS # refers to the free colupulone as there is no CAS # for the dicyclohexylammonium salt available.

Analysis Note

The counterion is determined quantitatively and treated as an impurity in the calculation of the absolute purity of this reference substance. The assigned absolute purity on the certificate of analysis of this reference standard, therefore, refers to the pure colupulone.

Other Notes

This compound is commonly found in plants of the genus: humulus

Legal Information

phyproof is a registered trademark of PhytoLab GmbH & Co. KG

Storage Class Code

11 - Combustible Solids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service