Skip to Content
Merck
All Photos(1)

Documents

G006032

HPLC Analysis MAb Variants on Proteomix® SCX-NP5, Fast Analysis

application for HPLC

Sign Into View Organizational & Contract Pricing


About This Item

technique(s)

HPLC: suitable

test parameters

column: Proteomix SCX-NP5, 25 cm x 4.6 mm I.D., 5 μm particles (Z777164)
mobile phase: [A] 2.4 mM Tris, 1.5 mM imidazole, 11.6 mM piperazine, pH 6.0; [B] A + 0.5 M NaCl, pH 10.5
gradient: top (0.8 mL/min): 5% B held for 5 min; to 29.5% B in 35 min; bottom (1.5 mL/min): 5% B held for 2.7 min; to 29.5% B in 18.6 min
flow rate: 0.8 mL/min or 1.5 mL/min
detector: UV 280 nm
injection: 20 μL
sample: MAb 321, 5 mg/mL

suitability

application for HPLC

application(s)

life science and biopharma

Analysis Note

Due to the low pressure of Proteomix SCX, faster analysis and high resolution can be achieved with a higher flow rate.

Legal Information

Proteomix is a registered trademark of Sepax Technologies

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service