Skip to Content
Merck
All Photos(1)

Documents

807475

TOYOPEARL® CM-650M Bulk Media

phase carboxymethyl, bottle of 250 mL, 65 μm particle size

Synonym(s):

TOYOPEARL® CM-650 Bulk Media

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
47101511

description

CM-650M

product line

TOYOPEARL®

form

slurry

feature

counter ion Na+

packaging

bottle of 250 mL

parameter

3 bar max. pressure

technique(s)

HPLC: suitable

matrix

methacrylate

matrix active group

carboxymethyl phase

particle size

65 μm

operating pH

5-10

capacity

0.10 meq/mL±0.02 meq/mL ion exchange capacity
40 g/L±10 adsorption capacity (lysozyme)
50 mg/mL±10 mg/mL binding capacity (bovine haemoglobin)

compatibility

mode of use weak cation exchange chromatography

separation technique

cation exchange

Looking for similar products? Visit Product Comparison Guide

General description

Toyopearl CM-650 resin is a weak cation exchanger for biomolecule purification. It can be used for high throughput capture, intermediate purification, and polishing process steps.

Application

TOYOPEARL® CM-650M is used in separation media and ion exchange resins. TOYOPEARL® CM-650M has been used in cloning studies.

Specifications

Clean in place with 0.5 M NaOH or 0.1 M HCl.

Physical form

Shipped in 20% (v/v) ethanol.

Recommended products

Discover LiChropur reagents ideal for HPLC or LC-MS analysis

Legal Information

Toyopearl is a registered trademark of Tosoh Corporation

Pictograms

Flame

Signal Word

Warning

Hazard Statements

Hazard Classifications

Flam. Liq. 3

WGK

WGK 1

Personal Protective Equipment

dust mask type N95 (US), Eyeshields, Gloves

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Don't see the Right Version?

If you require a particular version, you can look up a specific certificate by the Lot or Batch number.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Eri Shimizu et al.
Carbohydrate research, 338(24), 2841-2852 (2003-12-12)
An alginate lyase, termed HdAly in the present paper, was isolated from the hepatopancreas of abalone, Haliotis discus hannai, by ammonium sulfate fractionation, followed by TOYOPEARL CM-650M column chromatography. Enzymatic properties of HdAly were similar to those of previously reported
Ken-ichi Suzuki et al.
European journal of biochemistry, 270(4), 771-778 (2003-02-13)
A cellulase [endo-beta-1,4-D-glucanase (EC 3.2.1.4)] was isolated from the hepatopancreas of abalone Haliotis discus hannai by successive chromatographies on TOYOPEARL CM-650M, hydroxyapatite and Sephacryl S-200 HR. The molecular mass of the cellulase was estimated to be 66 000 Da by
Shuuji Ootsuka et al.
Journal of biotechnology, 125(2), 269-280 (2006-04-20)
An endo-beta-1,4-mannanase was isolated from digestive fluid of Pacific abalone, Haliotis discus hannai, by successive chromatographies on TOYPEARL CM-650M, hydroxyapatite, and TOYOPEARL HW50F. The abalone mannanase, named HdMan in the present paper, showed a molecular mass of approximately 39,000 Da

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service