Key Documents
55087-U
ValvTool
About This Item
UNSPSC Code:
41115854
NACRES:
SB.51
description
5/16 in. socket × 1/4 in. open wrench
General description
1/4
The ValvTool is a uniquely designed wrench that provides easy access for changing sample loops or replacing fittings on Rheodyne valves. The open end can be used for working with in. stainless steel HPLC fittings, as well.
Recommended products
Discover LiChropur reagents ideal for HPLC or LC-MS analysis
Choose from one of the most recent versions:
Certificates of Analysis (COA)