Skip to Content
Merck
All Photos(1)

Documents

33109-U

Supelco

Vials, crimp top, serum bottle

volume 50 mL, clear glass bottle, O.D. × H 43 mm × 73 mm, pkg of 272 ea

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41122001
NACRES:
NB.11

material

clear glass bottle

feature

autoclavable
closure type crimp top vial

packaging

pkg of 272 ea

O.D. × H

43 mm × 73 mm

volume

50 mL

fitting

thread for 20 mm

Looking for similar products? Visit Product Comparison Guide

Related Categories

General description

These vials are constructed of clear, borosilicate glass; closure not included.

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

Sorry, we don't have COAs for this product available online at this time.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service