Key Documents
Z741690
Sartorius low-retention pipette tips
volume range 2-120 μL, rack
About This Item
UNSPSC Code:
41121600
NACRES:
NB.01
Pricing and availability is not currently available.
material
colorless
sterility
sterile
feature
DNase free
RNase free
barrier
endotoxin free
low retention
packaging
pack of 960 ea (Racked)
manufacturer/tradename
Sartorius LH-LF790101
maximum volume
120 μL
volume range
2-120 μL
binding type
(liquid-repellent surface)
General description
Sartorius Low Retention Tips have an extremely even and durable liquid-repellent surface for optimising the flow of the liquid while dispensing, for optimum sample recovery. Liquids with low surface tension, like detergents, slide easily off of the inner surface of the tip, leaving very little residue behind. This Low Retention feature is thus very useful in the following applications, where such viscous, detergent containing, liquids are often pipetted: PCR, real-time PCR, cloning, SDS-PAGE and other nucleic acid or protein analysis methods. The Ttps are designed for best possible fitting and sealing to secure the highest possible accuracy and precision. These pipette tips enable ergonomic and light tip attachment by being compatible with the Optiload feature in Sartorius Picus, eLINE®, mLINE® and Proline® Plus pipettes. Tip trays are colour-coded for easy matching with corresponding, colour-coded Sartorius pipettes.
Other Notes
Browse all pipette tips
Legal Information
Proline is a registered trademark of Sartorius Biohit Liquid Handling Oy
eLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
mLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Choose from one of the most recent versions:
Certificates of Analysis (COA)