Skip to Content
Merck
All Photos(1)

Documents

Z729906

Hettich® fixed angle rotor, 48 place, 2 rows

for MIKRO 220/220R Centrifuge, Fixed Angle Rotor, 48 place, 2 rows (w/ bio-containment lid)

Synonym(s):

Hettich® centrifuge rotors

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41103900
NACRES:
NB.82

feature

RCF variable (21,255 outer row/18,845 inner row)
to hold 48 x 1.5/2.0 mL tubes

packaging

pack of 1 ea

manufacturer/tradename

Hettich 1158-L

parameter

21225 (18845) max. RCF (Inner (outer))

technique(s)

centrifugation: suitable

compatibility

for use with centrifuge models MIKRO 220/220R

Looking for similar products? Visit Product Comparison Guide

Related Categories

Legal Information

Hettich is a registered trademark of Andreas Hettich GmbH & Co. KG

Certificates of Analysis (COA)

Search for Certificates of Analysis (COA) by entering the products Lot/Batch Number. Lot and Batch Numbers can be found on a product’s label following the words ‘Lot’ or ‘Batch’.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service