Key Documents
Z709832
Sartorius eLINE® multichannel electronic pipettor
volume 50-1200 μL, 12 channel with USA plug
Synonym(s):
Biohit pipettes, electronic pipettes, pipettor
About This Item
UNSPSC Code:
41121500
Pricing and availability is not currently available.
AC/DC input
115 V AC
feature
electronic
manufacturer/tradename
Sartorius 730491
availability
not available in EU
volume
50-1200 μL
General description
With its easy-to-read display and simple-to-operate keyboard the hard work has been removed for programming Sartorius eLINE of the Biohitfamily. Boasting no fewer than 11 pipetting modes including:
  • Forward pipetting with optional cycle counting
  • Reverse and manual pipetting modes
  • Multi-dispensing; Diluting
  • Automated Dispensing
  • Mixing facility
The eLINE can store up to 6 user routines guaranteeing consistency of results irrelevant of the intricacy of the task undertaken. Tip ejection can be a chore at best and at worst a contributing factor to work-related upper limb disorders.
The eLINE negates this with its revolutionary electronic ejection system which discards the used tip at a touch of a button. Sartorius eLINE Electronic Pipettors should be charged using the original Sartorius eLINE Charging Stand. They also feature direct charging by using the Sartorius eLINE AC-adaptor.
Legal Information
eLINE is a registered trademark of Sartorius Biohit Liquid Handling Oy
Choose from one of the most recent versions:
Certificates of Analysis (COA)