Key Documents
Z659533
Bandelin SONOREX Ultrasonic bath accessories
tank lid to fit Z659517 and Z659509
About This Item
UNSPSC Code:
41103706
NACRES:
NB.74
Pricing and availability is not currently available.
material
stainless steel
manufacturer/tradename
Bandelin D 52
availability
not available in USA and Canada
General description
Stainless steel lid for an ultrasonic bath protects the bath liquid from external contamination and prevents unpleasant odors from the cleaning or disinfection agents added. In addition, the noise emitted when the ultrasonic cleaner is in operation, which may be perceived as annoying, is reduced. The lids are designed in such a way that any condensation that occurs is directed back into the bath.
Features and Benefits
  • protection of the bath fluid from contamination
  • sound-absorbing
  • shortens the heating phase
Legal Information
SONOREX is a trademark of Bandelin Electronic GmbH & Co. KG
Choose from one of the most recent versions:
Certificates of Analysis (COA)