Merck
All Photos(4)
Key Documents
Z616877
Vacuum controller for Buchi® evaporators and vacuum pumps
V-855, output230 V
About This Item
UNSPSC Code:
41104804
Pricing and availability is not currently available.
packaging
pack of 1 ea
manufacturer/tradename
Büchi 47298
availability
available only in EU
output
230 V
General description
Ideally suitable for the V-700/710 vacuum pumps, but is compatible with all older models. When used with the pump or the Rotavapor, a separate power cord is not required. However, if the vacuum controller is to be used as a stand-alone device it will require a power cord.
V-850 features:
  • Regulation to a specified setpoint
  • Library with 43 pre-determined solvents
  • Timer function
  • Control of jack and rotation on the Rotavapor
V-855 features:
  • EasyVac function when used with V-700/710 vacuum pump provides automatic process control based on vapor pressure detection
  • Automatic distillation with the unique stage probe
  • Programming for pressure gradients in specialized distillation
  • Repeat function for reproducible repetition of a distillation
Legal Information
Büchi is a registered trademark of Büchi Labortechnik AG
Choose from one of the most recent versions:
Certificates of Analysis (COA)