Skip to Content
Merck
All Photos(1)

Key Documents

Z379549

Cleaning supplies for camera equipment

Formula MC lens cleaner

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41100000
Pricing and availability is not currently available.

General description

Safe for cleaning and anti-fog protection of all glass and plastic lenses. Especially effective with the difficult job of cleaning multi-coated filters.
Dropper bottle contains 1 fluid ounce (approx. 28 mL).

Storage Class Code

10 - Combustible liquids

WGK

WGK 3

Flash Point(F)

Not applicable

Flash Point(C)

Not applicable


Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Questions

Reviews

No rating value

Active Filters

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service