Skip to Content
Merck
All Photos(1)

Key Documents

Z350397

Sigma-Aldrich

Carestream® BioMax® MR film

Synonym(s):

autoradiography film, xray film

Sign Into View Organizational & Contract Pricing


About This Item

UNSPSC Code:
41105341
NACRES:
NB.22

size

5 in. (13 cm) × 7 in. (18 cm)

Looking for similar products? Visit Product Comparison Guide

General description

Sheets of film with no interleaved paper
Single-emulsion film that provides maximum resolution and clearest background. The most sensitive film for direct 14C, 35S, and 33P autoradiography. Higher resolution than X-OMAT AR and BioMax MS for 32P direct and screen exposures and chemiluminescence, but with reduced sensitivity.

Legal Information

BioMax is a registered trademark of Carestream Health, Inc.
Carestream is a registered trademark of Carestream Health, Inc.
Kodak is a registered trademark of Eastman Kodak Company

Choose from one of the most recent versions:

Certificates of Analysis (COA)

Lot/Batch Number

It looks like we've run into a problem, but you can still download Certificates of Analysis from our Documents section.

If you need assistance, please contact Customer Support.

Already Own This Product?

Find documentation for the products that you have recently purchased in the Document Library.

Visit the Document Library

Marcos V Esteca et al.
International journal of molecular sciences, 21(21) (2020-11-01)
The high capacity of the skeletal muscle to regenerate is due to the presence of muscle stem cells (MuSCs, or satellite cells). The E3 ubiquitin ligase Parkin is a key regulator of mitophagy and is recruited to mitochondria during differentiation
Rastislav Horos et al.
Cell, 176(5), 1054-1067 (2019-02-19)
Vault RNAs (vtRNA) are small non-coding RNAs transcribed by RNA polymerase III found in many eukaryotes. Although they have been linked to drug resistance, apoptosis, and viral replication, their molecular functions remain unclear. Here, we show that vault RNAs directly
Shujuan Gao et al.
Journal of molecular signaling, 6, 8-8 (2011-08-13)
The family of A-kinase-anchoring proteins, AKAPs, constitutes a group of molecular scaffolds that act to catalyze dynamic interactions of protein kinase A, protein kinase C, tyrosine kinases, G-protein-coupled receptors and ion channels. AKAP5 (MW ~47 kDa) and AKAP12 (MW ~191
Atsuko Kimura et al.
The European journal of neuroscience, 30(2), 299-306 (2009-07-21)
The 5-HT(2C) receptor has been implicated in mood and eating disorders. In general, it is accepted that 5-HT(2C) receptor agonists increase anxiety behaviours and induce hypophagia. However, pharmacological analysis of the roles of these receptors is hampered by the lack

Articles

Background and protocols describing the various methods used by molecular biologists to detect samples of protein or nucleic acids bound to membranes.

Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.

Contact Technical Service