X4503
Anti-XIAP antibody produced in goat
affinity isolated antibody, lyophilized powder
Synonym(s):
Anti-HIAP2, Anti-X-linked Inhibitor of Apoptosis Protein
About This Item
biological source
goat
Quality Level
conjugate
unconjugated
antibody form
affinity isolated antibody
antibody product type
primary antibodies
clone
polyclonal
form
lyophilized powder
species reactivity
mouse, human, rat
technique(s)
western blot: 0.5 μg/mL using lysates from human Jurkat, mouse C2C12, or rat PC12 cells
UniProt accession no.
storage temp.
−20°C
target post-translational modification
unmodified
Gene Information
human ... BAZ1B(9031)
mouse ... Xiap(11798)
rat ... Xiap(63879)
Immunogen
Physical form
Disclaimer
Not finding the right product?
Try our Product Selector Tool.
Storage Class Code
11 - Combustible Solids
WGK
WGK 3
Flash Point(F)
Not applicable
Flash Point(C)
Not applicable
Personal Protective Equipment
Choose from one of the most recent versions:
Certificates of Analysis (COA)
Don't see the Right Version?
If you require a particular version, you can look up a specific certificate by the Lot or Batch number.
Already Own This Product?
Find documentation for the products that you have recently purchased in the Document Library.
Our team of scientists has experience in all areas of research including Life Science, Material Science, Chemical Synthesis, Chromatography, Analytical and many others.
Contact Technical Service